Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110778.1 | NP_001104248.1 | 12, 13 | 1268 | ||
NM_009613.2 | NP_033743.2 | 12, 13 | 1268 | |||
GenBank mRNA | AB009676.1 | - | 12, 13 | 1101 | ||
AK157926.1 | - | 12, 13 | 1273 | |||
BC054536.1 | - | 12, 13 | 1128 | 6400084 |