Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 8 | 1203 | ||
AK004712.1 | - | 4 | 209 | |||
AK128980.1 | - | 8 | 1279 | |||
AK158276.1 | - | 8 | 1227 | |||
AK159377.1 | - | 8 | 1197 | |||
AK159409.1 | - | 8 | 1227 | |||
AK159774.1 | - | 8 | 1198 | |||
AK159957.1 | - | 8 | 1198 | |||
AK196857.1 | - | 2 | 64 | |||
AK197169.1 | - | 2 | 64 | |||
AK204308.1 | - | 278 | ||||
BC082577.1 | - | 8 | 1215 | 30648530 |