Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053943.1 | - | 14 | 1842 | ||
AK004712.1 | - | 10 | 851 | |||
AK128980.1 | - | 14 | 1918 | |||
AK158276.1 | - | 14 | 1869 | |||
AK159330.1 | - | 14 | 1849 | |||
AK159342.1 | - | 14 | 1868 | |||
AK159377.1 | - | 14 | 1839 | |||
AK159409.1 | - | 14 | 1869 | |||
AK159774.1 | - | 14 | 1840 | |||
AK159957.1 | - | 14 | 1840 | |||
AK187128.1 | - | 2 | 107 | |||
BC082577.1 | - | 14 | 1854 | 30648530 | ||
X80478.1 | - | 5 | 572 |