Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 8 | 1415 | ||
GenBank mRNA | AB024935.1 | - | 8 | 1223 | ||
AK010636.1 | - | 8 | 1410 | |||
AK017742.1 | - | 8 | 1232 | |||
AK075932.1 | - | 8 | 1282 | |||
AK075987.1 | - | 8 | 1326 | |||
AK077538.1 | - | 8 | 1416 | |||
AK088321.1 | - | 1 | 497 | |||
AK167443.1 | - | 8 | 1389 | |||
AK168073.1 | - | 8 | 1325 | |||
AK168210.1 | - | 8 | 1325 | |||
AK168721.1 | - | 8 | 1265 | |||
AY281130.1 | - | 8 | 1442 | |||
BC085264.1 | - | 8 | 1312 | 6392001 |