Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019805.4 | NP_062779.3 | 4 | 571 | ||
GenBank mRNA | AF076607.2 | - | 4 | 535 | ||
AK030775.1 | - | 4 | 536 | |||
AK049968.1 | - | 4 | 510 | |||
AK137286.1 | - | 4 | 535 | |||
AK150764.1 | - | 4 | 535 | |||
AK150891.1 | - | 4 | 535 | |||
AK153114.1 | - | 4 | 535 | |||
AK159523.1 | - | 4 | 503 | |||
AK164202.1 | - | 4 | 540 | |||
AK188802.1 | - | 1 | 38 | |||
BC006635.1 | - | 4 | 524 | 3482261 |