Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019814.4 | NP_062788.1 | 3 | 634 | ||
GenBank mRNA | AB253941.1 | - | 2 | 146 | ||
AF141312.1 | - | 3 | 165 | |||
AK004166.1 | - | 3 | 261 | |||
AK008756.1 | - | 3 | 261 | |||
AK018360.1 | - | 3 | 262 | |||
AY028386.1 | - | 3 | 187 | |||
AY028387.1 | - | 3 | 172 | |||
BC021594.1 | - | 3 | 184 | 5148784 | ||
BC090259.1 | - | 3 | 238 | 5060413 | ||
BC132371.1 | - | 3 | 252 | 40130648 | ||
BC151191.1 | - | 3 | 252 | 8861324 | ||
BE457620.1 | - | 3 | 430 |