Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020271.3 | NP_064667.2 | 2 | 1416 | ||
GenBank mRNA | AB041662.1 | - | 2 | 1312 | ||
AK043228.1 | - | 2 | 1377 | |||
AY366300.1 | - | 2 | 1378 | |||
BC037010.1 | - | 2 | 840 | |||
BC046955.1 | - | 2 | 1367 | |||
BC049107.1 | - | 2 | 1416 | |||
BC058388.1 | - | 2 | 962 | |||
BC069982.1 | - | 2 | 1188 |