Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023126.2 | NP_075615.2 | 8 | 1916 | ||
GenBank mRNA | AK076048.1 | - | 8 | 1795 | ||
AK079306.1 | - | 1783 | ||||
AK080740.1 | - | 8 | 1768 | |||
BY700504.1 | - | 1 | 298 |