Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023554.2 | NP_076043.2 | 6, 7 | 636 | ||
GenBank mRNA | AB041566.1 | - | 6, 7 | 620 | ||
AK008670.1 | - | 6, 7 | 621 | |||
AK019972.1 | - | 6, 7 | 621 | |||
AK032963.1 | - | 6, 7 | 622 | |||
BC090637.1 | - | 6, 7 | 609 | 30940523 | ||
BC130012.1 | - | 6, 7 | 626 | 40140312 |