Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023554.2 | NP_076043.2 | 8 | 1591 | ||
GenBank mRNA | AB041566.1 | - | 8 | 1575 | ||
AK032963.1 | - | 8 | 1577 | |||
AK140839.1 | - | 583 | ||||
BC090637.1 | - | 8 | 1564 | 30940523 |