Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023598.2 | NP_076087.2 | 2 | 276 | ||
GenBank mRNA | AF169968.1 | - | 501 | |||
AF280065.1 | - | 2 | 290 | |||
AK017396.1 | - | 2 | 277 | |||
AK029818.1 | - | 2 | 272 | |||
AK031596.1 | - | 2 | 272 | |||
BY717211.1 | - | 2 | 277 |