Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003681.2 | NP_001003681.1 | 3 | 314 | ||
NR_027780.1 | 3 | 314 | ||||
XM_006724100.3 | XP_006724163.1 | 2 | 393 | |||
XM_006724101.3 | XP_006724164.1 | 2 | 393 | |||
XM_006724102.1 | XP_006724165.1 | 2 | 182 | |||
GenBank mRNA | AJ010070.1 | - | 3 | 263 | ||
AK299833.1 | - | 3 | 279 | |||
AL079310.1 | - | 3 | 263 | |||
BC017326.2 | - | 2 | 151 | |||
BC025303.2 | - | 2 | 209 | |||
BC052624.1 | - | 3 | 268 | |||
BC062413.1 | - | 3 | 266 | |||
BC080611.1 | - | 3 | 269 | |||
BC111373.1 | - | 2 | 218 | |||
CB991207.1 | - | 3 | 311 | |||
CR456504.1 | - | 2 | 170 | |||
DW452042.1 | - | 2 | 232 |