Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023645.3 | NP_076134.1 | 2 | 522 | ||
GenBank mRNA | AJ404004.1 | - | 2 | 284 | ||
AK007835.1 | - | 2 | 515 | |||
AK035391.1 | - | 2 | 515 | |||
AK035899.1 | - | 2 | 523 | |||
AK053315.1 | - | 2 | 515 | |||
AK077468.1 | - | 1 | 1625 | |||
AK079701.1 | - | 2 | 515 | |||
AK170300.1 | - | 2 | 551 | |||
BC023141.1 | - | 2 | 502 | 4018698 |