Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024206.4 | NP_077168.2 | 9 | 1180 | ||
GenBank mRNA | AK003354.1 | - | 9 | 1181 | ||
AK009755.1 | - | 8 | 1164 | |||
BC002128.1 | - | 9 | 1174 | 3590767 |