Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024432.2 | NP_077752.1 | 11 | 1665 | ||
GenBank mRNA | AK008942.1 | - | 11 | 1673 | ||
AK009117.1 | - | 10 | 1447 | |||
AK075705.1 | - | 9 | 2729 | |||
AK082510.1 | - | 1705 | ||||
AK131187.1 | - | 5 | 3437 | |||
AK148170.1 | - | 1633 | ||||
AK161273.1 | - | 11 | 1687 | |||
AK179041.1 | - | 1 | 103 | |||
AK188888.1 | - | 1 | 103 | |||
AK200815.1 | - | 230 | ||||
AK220061.1 | - | 1 | 352 | |||
BC049963.1 | - | 11 | 1665 | 2655935 |