Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145713.1 | NP_001139185.1 | 2 | 291 | ||
NM_001145714.1 | NP_001139186.1 | 2 | 291 | |||
NM_005500.2 | NP_005491.1 | 2 | 291 | |||
NR_027280.1 | 2 | 340 | ||||
XM_006722963.2 | XP_006723026.1 | 2 | 291 | |||
XM_006722965.3 | XP_006723028.1 | 2 | 291 | |||
XM_017026135.1 | XP_016881624.1 | 2 | 291 | |||
XM_017026136.1 | XP_016881625.1 | 2 | 162 | |||
GenBank mRNA | AF046025.2 | - | 2 | 228 | ||
AF090385.2 | - | 2 | 236 | |||
AF110956.1 | - | 2 | 183 | |||
AF161489.1 | - | 2 | 239 | |||
AK021978.1 | - | 2 | 285 | |||
AK027595.1 | - | 2 | 255 | |||
AK055481.1 | - | 2 | 256 | |||
AK090645.1 | - | 2 | 309 | |||
AK302287.1 | - | 2 | 109 | |||
AK315624.1 | - | 2 | 233 | |||
BC000344.2 | - | 2 | 244 | 2823028 | ||
BC003611.2 | - | 2 | 212 | 2988252 | ||
BC018271.1 | - | 2 | 251 | 3452340 | ||
BT007290.1 | - | 2 | 183 | |||
DA047601.1 | - | 2 | 296 | |||
DA086973.1 | - | 2 | 292 | |||
DC297573.1 | - | 2 | 291 | |||
DC312737.1 | - | 2 | 340 |
Gene Symbol: LOC341056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027288.1 | 1 | 257 |