Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025970.3 | NP_080246.1 | 3 | 179 | ||
GenBank mRNA | AK007377.1 | - | 3 | 169 | ||
AK008007.1 | - | 3 | 180 | |||
AK132279.1 | - | 190 | ||||
AY071852.1 | - | 3 | 168 | |||
BC026442.1 | - | 174 | ||||
BC037657.1 | - | 149 | ||||
BC046825.1 | - | 186 | ||||
BC094629.1 | - | 3 | 210 | 3376726 | ||
BC125606.1 | - | 3 | 180 | 40130121 | ||
BC125608.1 | - | 3 | 180 | 40130123 |