Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026474.5 | NP_080750.1 | 10 | 727 | ||
GenBank mRNA | AK007881.1 | - | 10 | 662 | ||
AK010711.1 | - | 9 | 696 | |||
AK010830.1 | - | 10 | 644 | |||
AK020433.1 | - | 10 | 646 | |||
AK045153.1 | - | 10 | 666 | |||
AK082328.1 | - | 10 | 729 | |||
AK141416.1 | - | 663 | ||||
AK148838.1 | - | 10 | 641 | |||
BC009167.1 | - | 10 | 644 | 2811924 |