Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026474.5 | NP_080750.1 | 13 | 1731 | ||
GenBank mRNA | AK082328.1 | - | 13 | 1733 | ||
AK141416.1 | - | 1667 |