Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005527.3 | NP_005518.3 | 2 | 424 | ||
GenBank mRNA | AK223386.1 | - | 2 | 398 | ||
AK295416.1 | - | 2 | 658 | |||
AK302179.1 | - | 2 | 393 | |||
AK315227.1 | - | 2 | 334 | |||
BC034483.1 | - | 2 | 401 | 4824979 | ||
BG717447.1 | - | 2 | 437 | |||
D85730.1 | - | 2 | 376 |