Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026817.3 | NP_081093.2 | 6 | 482 | ||
GenBank mRNA | AK004012.1 | - | 6 | 428 | ||
AK170205.1 | - | 3 | 235 | |||
BC030376.1 | - | 6 | 449 | 5374611 | ||
BQ887131.1 | - | 6 | 474 |