Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 9 | 822 | ||
NM_001287211.1 | NP_001274140.1 | 10 | 885 | |||
NM_026972.5 | NP_081248.4 | 10 | 912 | |||
GenBank mRNA | AF373409.1 | - | 10 | 890 | ||
AF374471.1 | - | 9 | 779 | |||
AF422108.1 | - | 10 | 885 | |||
AF424790.1 | - | 10 | 884 | |||
AF424791.1 | - | 10 | 884 | |||
AF424792.1 | - | 10 | 884 | |||
AF424793.1 | - | 10 | 884 | |||
AF424794.1 | - | 10 | 904 | |||
AF424795.1 | - | 10 | 901 | |||
AF424796.1 | - | 10 | 895 | |||
AF424797.1 | - | 9 | 794 | |||
AF424798.1 | - | 9 | 814 | |||
AF424799.1 | - | 10 | 857 | |||
AF424800.1 | - | 4 | 315 | |||
AF424801.1 | - | 4 | 315 | |||
AF424802.1 | - | 4 | 315 | |||
AF424803.1 | - | 9 | 768 | |||
AK007656.1 | - | 9 | 828 | |||
AK138999.1 | - | 10 | 886 |