Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190453.1 | NP_001177382.1 | 6 | 551 | ||
NM_001190454.1 | NP_001177383.1 | 5 | 560 | |||
NM_027151.2 | NP_081427.1 | 5 | 545 | |||
GenBank mRNA | AK144464.1 | - | 5 | 421 | ||
AK144505.1 | - | 5 | 421 | |||
AK164021.1 | - | 5 | 422 | |||
BC004613.1 | - | 5 | 408 | 3585310 |