Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027629.3 | NP_081905.1 | 7 | 1095 | ||
GenBank mRNA | AK016437.1 | - | 5 | 454 | ||
AK039189.1 | - | 7 | 1079 | |||
BC020351.1 | - | 7 | 1308 | |||
BC032150.1 | - | 3 | 301 | 5004308 | ||
BC076571.1 | - | 7 | 992 | 30356163 |