Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027759.3 | NP_082035.1 | 3, 4 | 482 | ||
GenBank mRNA | AF448787.1 | - | 3, 4 | 482 | ||
AK005924.1 | - | 3, 4 | 473 | |||
AK017026.1 | - | 3, 4 | 471 | |||
BC125024.1 | - | 3, 4 | 374 | 40056533 |