Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029988.2 | NP_084264.1 | 1, 2 | 229 | ||
GenBank mRNA | AK020902.1 | - | 1, 2 | 205 | ||
AK030744.1 | - | 1, 2 | 193 | |||
AK167892.1 | - | 1, 2 | 211 | |||
BC086806.1 | - | 1, 2 | 180 | 30603877 | ||
BY737882.1 | - | 1, 2 | 211 | |||
BY779412.1 | - | 1, 2 | 229 |