Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029988.2 | NP_084264.1 | 4 | 619 | ||
GenBank mRNA | AK030744.1 | - | 3 | 1702 | ||
AK167892.1 | - | 4 | 601 | |||
BC086806.1 | - | 4 | 570 | 30603877 | ||
BQ963876.1 | - | 2 | 178 | |||
BY737882.1 | - | 4 | 601 |