Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204134.1 | NP_001191063.1 | 6 | 1111 | ||
NM_030888.4 | NP_112150.1 | 6 | 917 | |||
GenBank mRNA | AF246265.1 | - | 6 | 894 | ||
AK029352.1 | - | 6 | 894 | |||
AK075867.1 | - | 6 | 894 | |||
AK081704.1 | - | 6 | 894 | |||
AK128898.1 | - | 6 | 917 | |||
BC019636.1 | - | 6 | 1120 | |||
BC055047.1 | - | 6 | 879 | 6486507 | ||
BC055859.1 | - | 6 | 858 | 4238617 |