Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039087.1 | NP_001034176.1 | 3 | 549 | ||
NM_054050.2 | NP_473391.1 | 3 | 549 | |||
GenBank mRNA | AF348669.1 | - | 3 | 505 | ||
AK147468.1 | - | 3 | 550 | |||
AK147470.1 | - | 3 | 550 | |||
AK147506.1 | - | 3 | 518 |