Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291320.1 | NP_001278249.1 | 11 | 1396 | ||
NM_133709.3 | NP_598470.3 | 11 | 1438 | |||
GenBank mRNA | AK007577.1 | - | 11 | 1401 | ||
BC019399.1 | - | 11 | 1403 | 4012143 |