Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133726.2 | NP_598487.1 | 3 | 332 | ||
GenBank mRNA | AK013981.1 | - | 3 | 319 | ||
AK146528.1 | - | 3 | 328 | |||
AK147169.1 | - | 355 | ||||
AK150686.1 | - | 303 | ||||
AK151631.1 | - | 303 | ||||
AK152270.1 | - | 3 | 328 | |||
AK154658.1 | - | 321 | ||||
AK159149.1 | - | 312 | ||||
AK205921.1 | - | 2 | 89 | |||
BC003843.1 | - | 3 | 332 | 3495262 | ||
CT010318.1 | - | 3 | 232 |