Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133779.2 | NP_598540.2 | 12 | 1562 | ||
GenBank mRNA | AK019717.2 | - | 12 | 1552 | ||
AK044499.1 | - | 12 | 1546 | |||
AK049171.1 | - | 12 | 1556 | |||
AK153462.1 | - | 12 | 1548 | |||
AK153464.1 | - | 12 | 1548 | |||
AK155057.1 | - | 12 | 1549 | |||
AK157226.1 | - | 12 | 1542 | |||
BC016520.1 | - | 13 | 1616 | |||
BC034175.1 | - | 12 | 1538 | 5026752 |