Search Thermo Fisher Scientific
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133823.4 | NP_598584.2 | 6, 7 | 1052 | ||
GenBank mRNA | AK050255.1 | - | 6, 7 | 1034 | ||
AK160235.1 | - | 7, 8 | 1379 | |||
BC021954.1 | - | 5, 6 | 1003 | 5134853 | ||
BC027398.1 | - | 4, 5 | 463 | |||
BC085229.1 | - | 5, 6 | 1011 |