Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170430.1 | NP_001163901.1 | 4, 5 | 701 | ||
NM_001170431.1 | NP_001163902.1 | 4, 5 | 836 | |||
NM_134060.4 | NP_598821.1 | 4, 5 | 679 | |||
GenBank mRNA | AK076592.1 | - | 4, 5 | 674 | ||
AK077208.1 | - | 4, 5 | 655 | |||
AK141280.1 | - | 4, 5 | 837 | |||
BC006881.1 | - | 4, 5 | 693 | 3596363 | ||
CX238927.1 | - | 3, 4 | 385 |