Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290574.1 | NP_001277503.1 | 3, 4 | 1178 | ||
NM_138605.3 | NP_613071.3 | 3, 4 | 1175 | |||
GenBank mRNA | AF229644.1 | - | 3, 4 | 1014 | ||
BC059275.1 | - | 3, 4 | 1112 | |||
BC138137.1 | - | 3, 4 | 1007 | 8861156 | ||
BC138138.1 | - | 3, 4 | 1007 | 8861157 |