Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145467.2 | NP_663442.2 | 10, 11 | 1570 | ||
GenBank mRNA | AK029068.1 | - | 10, 11 | 1537 | ||
AK047014.1 | - | 1029 | ||||
AK051773.1 | - | 1608 | ||||
BC020152.1 | - | 10, 11 | 1537 | 3708340 |