Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172298.2 | NP_758502.1 | 2 | 361 | ||
GenBank mRNA | AY063491.1 | - | 2 | 361 | ||
BC150674.1 | - | 2 | 304 | 9087585 |