Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145713.1 | NP_001139185.1 | 2 | 300 | ||
NM_001145714.1 | NP_001139186.1 | 2 | 300 | |||
NM_005500.2 | NP_005491.1 | 2 | 300 | |||
NR_027280.1 | 2 | 349 | ||||
XM_006722963.2 | XP_006723026.1 | 2 | 300 | |||
XM_006722965.3 | XP_006723028.1 | 2 | 300 | |||
XM_017026135.1 | XP_016881624.1 | 2 | 300 | |||
XM_017026136.1 | XP_016881625.1 | 2 | 171 | |||
GenBank mRNA | AF046025.2 | - | 2 | 237 | ||
AF090385.2 | - | 2 | 245 | |||
AF110956.1 | - | 2 | 192 | |||
AF161489.1 | - | 2 | 248 | |||
AK021978.1 | - | 2 | 294 | |||
AK027595.1 | - | 2 | 264 | |||
AK055481.1 | - | 2 | 265 | |||
AK090645.1 | - | 2 | 318 | |||
AK302287.1 | - | 2 | 118 | |||
AK315624.1 | - | 2 | 242 | |||
BC000344.2 | - | 2 | 253 | 2823028 | ||
BC003611.2 | - | 2 | 221 | 2988252 | ||
BC018271.1 | - | 2 | 260 | 3452340 | ||
BT007290.1 | - | 2 | 192 | |||
DA047601.1 | - | 2 | 305 | |||
DA086973.1 | - | 2 | 301 | |||
DC297573.1 | - | 2 | 300 | |||
DC312737.1 | - | 2 | 349 |
Gene Symbol: LOC341056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027288.1 | 1 | 266 |