Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291003.1 | NP_001277932.1 | 3, 4 | 314 | ||
NM_172539.3 | NP_766127.1 | 3, 4 | 445 | |||
GenBank mRNA | AK033037.1 | - | 3, 4 | 445 | ||
BC064729.1 | - | 3, 4 | 306 | 30476764 | ||
DV649054.1 | - | 3, 4 | 326 | |||
FJ187790.1 | - | 3, 4 | 253 | |||
FJ187791.1 | - | 2, 3 | 190 |