Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291049.1 | NP_001277978.1 | 15 | 4170 | ||
NM_173754.4 | NP_776115.2 | 15 | 4185 | |||
GenBank mRNA | AK083271.1 | - | 15 | 4023 | ||
AK083574.1 | - | 1 | 481 | |||
BC008156.1 | - | 1 | 1100 |