Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177322.3 | NP_796296.1 | 3 | 2048 | ||
GenBank mRNA | BC036175.1 | - | 3 | 1946 | 4989471 |
Set Membership: |
G-protein coupled receptor |