Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178680.4 | NP_848795.3 | 4 | 350 | ||
GenBank mRNA | AF539793.1 | - | 3 | 243 | ||
AK052294.1 | - | 4 | 277 | |||
AK081821.1 | - | 4 | 289 | |||
AK084418.1 | - | 4 | 318 | |||
AK168968.1 | - | 4 | 318 | |||
AK169274.1 | - | 4 | 289 | |||
BC084585.1 | - | 4 | 283 | 30429260 | ||
BY133112.1 | - | 4 | 350 |