Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178680.4 | NP_848795.3 | 20 | 3328 | ||
GenBank mRNA | AF539793.1 | - | 19 | 3227 | ||
AK052294.1 | - | 20 | 3255 | |||
BC084585.1 | - | 20 | 3261 | 30429260 |