Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp385b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178723.6 | NP_848838.2 | 3 | 618 | ||
GenBank mRNA | AK033075.1 | - | 3 | 579 | ||
AK046798.1 | - | 3 | 619 | |||
AK079322.1 | - | 3 | 500 | |||
AK149352.1 | - | 3 | 599 | |||
BC132352.1 | - | 1 | 197 | 40130629 | ||
BC138239.1 | - | 1 | 197 | 8861258 |