Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181579.1 | NP_853557.1 | 2, 3 | 413 | ||
GenBank mRNA | AF309775.1 | - | 1, 2 | 306 | ||
AK036922.1 | - | 2, 3 | 418 | |||
AK086511.1 | - | 2, 3 | 414 | |||
AK137653.1 | - | 2, 3 | 414 |