Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182785.3 | NP_877586.1 | 4 | 513 | ||
GenBank mRNA | AK077071.1 | - | 4 | 514 | ||
BC049744.1 | - | 3 | 313 | 6772266 |