Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182939.4 | NP_891984.1 | 6, 7 | 748 | ||
GenBank mRNA | AK082539.1 | - | 6, 7 | 749 | ||
AK084169.1 | - | 6, 7 | 709 | |||
BC038014.1 | - | 3, 4 | 177 | |||
BC056340.1 | - | 6, 7 | 568 | 5703151 | ||
BC062117.1 | - | 5, 6 | 422 | |||
BC098464.1 | - | 6, 7 | 514 | 30093511 | ||
BC110429.1 | - | 6, 7 | 753 | 5350332 |