Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182939.4 | NP_891984.1 | 8, 9 | 1184 | ||
GenBank mRNA | AK082539.1 | - | 8, 9 | 1185 | ||
AK084169.1 | - | 8, 9 | 1145 | |||
BC030412.1 | - | 2, 3 | 351 | |||
BC038014.1 | - | 5, 6 | 613 | |||
BC056340.1 | - | 8, 9 | 1004 | 5703151 | ||
BC062117.1 | - | 7, 8 | 858 | |||
BC098464.1 | - | 8, 9 | 947 | 30093511 | ||
BC110429.1 | - | 8, 9 | 1189 | 5350332 | ||
BC112399.1 | - | 6, 7 | 618 |