Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nlrp9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042612.1 | NP_001036077.1 | 2 | 1739 | ||
GenBank mRNA | AK145425.1 | - | 1 | 1352 | ||
AK163321.1 | - | 2 | 1508 | |||
AY360472.1 | - | 1734 | ||||
AY596203.1 | - | 2 | 1739 |