Gene Symbol: Rnf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194346.2 | NP_919327.2 | 10 | 1857 | ||
GenBank mRNA | AK131160.1 | - | 4 | 1637 | ||
AK156320.1 | - | 10 | 1940 | |||
AK157962.1 | - | 9 | 2147 | |||
AY283085.1 | - | 10 | 1766 | |||
BC031509.1 | - | 8 | 1798 | |||
BC057595.1 | - | 10 | 1857 | 6811051 |